4-Brom-3-(methylsulfonyl)benzotrifluorid

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-2-(methylsulphonyl)-4-(trifluoromethyl)benzene

OT-1175-5G OT-1175-1G
COBBOT-1175-5GEA 595 EUR
COBBOT-1175-5G COBBOT-1175-1G
4-Brom-3-(methylsulfonyl)benzotrifluorid
4-Brom-3-(methylsulfonyl)benzotrifluorid
Formel: C₈H₆BrF₃O₂S
Molecular Weight: 303,096 g/mol
MDL: MFCD03094286
CAS-Nummer: 1215205-98-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR