2-Brom-1-methyl-4-(trifluormethylsulfonyl)benzol

Lieferant: COMBI-BLOCKS

OT-0822-1G OT-0822-5G
COBBOT-0822-1GEA 451 EUR
COBBOT-0822-1G COBBOT-0822-5G
2-Brom-1-methyl-4-(trifluormethylsulfonyl)benzol
2-Brom-1-methyl-4-(trifluormethylsulfonyl)benzol
Formel: C₈H₆BrF₃O₂S
MDL: MFCD11855954
CAS-Nummer: 1133116-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR