3-Brom-5-(methylsulfonyl)benzotrifluorid
Lieferant: COMBI-BLOCKS
Synonyme:
1-Bromo-3-(methylsulphonyl)-5-(trifluoromethyl)benzene
OT-1005-5G
OT-1005-1G
COBBOT-1005-5GEA
864
EUR
COBBOT-1005-5G
COBBOT-1005-1G
3-Brom-5-(methylsulfonyl)benzotrifluorid
3-Brom-5-(methylsulfonyl)benzotrifluorid
Formel:
C₈H₆BrF₃O₂S |
MDL:
MFCD15143584 CAS-Nummer: 1215205-96-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...