Indol-4-carbonitril ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H63391.03
H63391.06
H63391.03EA
34.5
EUR
H63391.03
H63391.06
Indol-4-carbonitril ≥99%
Indol-4-carbonitril
Purity: 99%
CAS No.: 16136-52-0
Molecular Formula: C9H6N2
Formula Weight: 142.16
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 120-125°
MDL No.: MFCD00152045
Formel:
C₉H₆N₂ Schmelzpunkt: 120…125 °C |
MDL:
MFCD00152045 CAS-Nummer: 16136-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...