3-Methyladenin
Lieferant: Cayman Chemical
Synonyme:
3-Methyl-3H-adenin, 6-Amino-3-methylpurin
13242-25
13242-50
13242-100
CAYM13242-25EA
44
EUR
CAYM13242-25
CAYM13242-50
CAYM13242-100
3-Methyladenin
3-Methyladenin
Formel:
C₆H₇N₅ Molecular Weight: 149,16 g/mol |
MDL:
MFCD00010531 CAS-Nummer: 5142-23-4 EINECS: 225-908-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...