Ammoniumcer(IV)nitrat 0.01 mol/l (0.01 N) in wässriger Lösung Reag. Ph. Eur.

Lieferant: Bernd Kraft
Achtung

Synonyme: Diammoniumhexanitratocerat, di-Ammonium-hexanitratocerat (IV), Cer(IV)ammoniumnitrat

25001.3100 25001.4100 25001.5400
KRAF25001.3100EA 233 EUR
KRAF25001.3100 KRAF25001.4100 KRAF25001.5400
Ammoniumcer(IV)nitrat 0.01 mol/l (0.01 N) in wässriger Lösung Reag. Ph. Eur.
Ammoniumcer(IV)nitrat
Formel: (NH₄)₂Ce(NO₃)₆
Molecular Weight: 548,22 g/mol
MDL: MFCD00151121
CAS-Nummer: 16774-21-3
EINECS: 240-827-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR