3-Brom-2-fluoranisol

Lieferant: COMBI-BLOCKS

Synonyme: 1-Bromo-2-fluoro-3-methoxybenzene, 3-Bromo-2-fluorophenyl methyl ether

OT-2094-1G OT-2094-5G
COBBOT-2094-1GEA 90.2 EUR
COBBOT-2094-1G COBBOT-2094-5G
3-Brom-2-fluoranisol
3-Brom-2-fluoranisol
Formel: C₇H₆BrFO
Molecular Weight: 205.03 g/mol
MDL: MFCD07780183
CAS-Nummer: 295376-21-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR