4-Fluor-2-nitroanisol ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Fluoro-1-methoxy-2-nitrobenzene

H61110.06 H61110.14
H61110.06EA 39.1 EUR
H61110.06 H61110.14
4-Fluor-2-nitroanisol ≥98%
4-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
Molecular Weight: 171,13 g/mol
Schmelzpunkt: 62…64 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00013375
CAS-Nummer: 445-83-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR