2-Amino-6-methyl-3-nitropyridin 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 6-Methyl-3-nitropyridin-2-amin, 2-Amino-3-nitro-6-picolin, 6-Methyl-3-nitro-2-pyridylamin

H61729.06 H61729.14
H61729.06EA 35.3 EUR
H61729.06 H61729.14
2-Amino-6-methyl-3-nitropyridin 98%
2-Amino-6-methyl-3-nitropyridin
Formel: C₆H₇N₃O₂
Molecular Weight: 153,14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047443
CAS-Nummer: 21901-29-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR