3,4-Dichlorpyridin ≥98%

Lieferant: Thermo Scientific
Achtung

H63749.03 H63749.06
H63749.03EA 36.1 EUR
H63749.03 H63749.06
3,4-Dichlorpyridin ≥98%
3,4-Dichloropyridine

Purity: 98% CAS No.: 55934-00-4 Molecular Formula: C5H3Cl2N Formula Weight: 147.99 Physical Form: Liquid Appearance: Colourless UN No.: 2810 MDL No.: MFCD01861989

Formel: C₅H₃Cl₂N
Siedepunkt: 182…183 °C
Schmelzpunkt: 22…24 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01861989
CAS-Nummer: 55934-00-4
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR