5-Amino-2-methylpyridin

Lieferant: COMBI-BLOCKS

Synonyme: 6-Methylpyridin-3-amin, 3-Amino-6-picolin, 6-Methyl-3-pyridylamin, 5-Amino-2-picolin

PY-7900-1G PY-7900-25G PY-7900-5G
COBBPY-7900-1GEA 25.5 EUR
COBBPY-7900-1G COBBPY-7900-25G COBBPY-7900-5G
5-Amino-2-methylpyridin
5-Amino-2-methylpyridin
Formel: C₆H₈N₂
Molecular Weight: 108.14 g/mol
MDL: MFCD00833389
CAS-Nummer: 3430-14-6
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR