2-Fluormesitylen

Lieferant: Thermo Scientific

Synonyme: 2,4,6-Trimethylfluorbenzol

HAN00038.1 HAN00038.10
MAYBHAN00038.1EA 0 EUR
MAYBHAN00038.1 MAYBHAN00038.10
2-Fluormesitylen
2-Fluormesitylen
Formel: C₉H₁₁F
Molecular Weight: 138,18 g/mol
MDL: MFCD00052365
CAS-Nummer: 392-69-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR