2-Chlor-N-methylanilin

Lieferant: Thermo Scientific

TL00240.1 TL00240.10 TL00240.50
MAYBTL00240.1EA 0 EUR
MAYBTL00240.1 MAYBTL00240.10 MAYBTL00240.50
2-Chlor-N-methylanilin
2-Chlor-N-methylanilin
Formel: C₇H₈ClN
Molecular Weight: 141,6 g/mol
Siedepunkt: 95…96 °C (20 mmHg)
Dichte: 1,15 g/cm³ (20 °C)
MDL: MFCD00045170
CAS-Nummer: 932-32-1
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR