3-Chlor-N-methylanilin

Lieferant: Thermo Scientific

Synonyme: N-Methyl-3-chloroaniline

TL00250.1 TL00250EA TL00250EE
MAYBTL00250.1EA 0 EUR
MAYBTL00250.1 MAYBTL00250EA MAYBTL00250EE
3-Chlor-N-methylanilin
3-Chlor-N-methylanilin
Formel: C₇H₈ClN
Molecular Weight: 141,6 g/mol
Siedepunkt: 235 °C (1013 hPa)
Dichte: 1,174 g/cm³ (11.5 °C)
MDL: MFCD00052014
CAS-Nummer: 7006-52-2
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR