1-Benzyl-3-methylimidazoliumchlorid ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 1-Benzyl-3-methyl-1H-imidazol-3-ium chloride

H59997.06 H59997.18
H59997.06EA 43 EUR
H59997.06 H59997.18
1-Benzyl-3-methylimidazoliumchlorid ≥97%
1-Benzyl-3-methylimidazoliumchlorid
Formel: C₁₁H₁₃ClN₂
Molecular Weight: 208.69 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07784451
CAS-Nummer: 36443-80-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR