5-Nitro-8-chinolinol
Lieferant: COMBI-BLOCKS
Synonyme:
Nitroxolin
PY-7805-100G
PY-7805-25G
PY-7805-5G
COBBPY-7805-100GEA
0
EUR
COBBPY-7805-100G
COBBPY-7805-25G
COBBPY-7805-5G
5-Nitro-8-chinolinol
5-Nitro-8-chinolinol
Formel:
C₉H₆N₂O₃ Molecular Weight: 190,16 g/mol Siedepunkt: 419 °C (1013 hPa) Schmelzpunkt: 179…181 °C Dichte: 1,549 g/cm³ (22 °C) |
MDL:
MFCD00006791 CAS-Nummer: 4008-48-4 EINECS: 223-662-4 Merck Index: 12,06753 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...