Zimmermann-Reinhardt-Lösung
Lieferant: Bernd Kraft
|
Gefahr
|
05465.2000
05465.3000
KRAF05465.2000EA
0
EUR
KRAF05465.2000
KRAF05465.3000
Zimmermann-Reinhardt-Lösung
Zimmermann-Reinhardt-Lösung
Molecular Weight:
223,06 g/mol |
UN:
1760 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...