4'-Brom-2'-hydroxyacetophenon 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H61137.03
H61137.06
H61137.03EA
109
EUR
H61137.03
H61137.06
4'-Brom-2'-hydroxyacetophenon 95%
4'-Brom-2'-hydroxyacetophenon
Formel:
C₈H₇BrO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD03428533 CAS-Nummer: 30186-18-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...