2-Brom-4-(trifluormethyl)phenol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Bromo-4-hydroxybenzotrifluoride
H61836.03
H61836.06
H61836.03EA
46
EUR
H61836.03
H61836.06
2-Brom-4-(trifluormethyl)phenol ≥98%
2-Brom-4-(trifluormethyl)phenol
Purity: 98%
CAS No.: 81107-97-3
Molecular Formula: C7H4BrF3O
Formula Weight: 241.01
Physical Form: Liquid
Appearance: Brown
Beilstein Reference: 7702274
MDL No.: MFCD06657954
Formel:
C₇H₄BrF₃O Molecular Weight: 241.01 Lagertemperatur: Raumtemperatur |
MDL:
MFCD06657954 CAS-Nummer: 81107-97-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...