2-Brom-4-nitroimidazol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Brom--5-nitro1H-imidazol
H61155.03
H61155.06
H61155.03EA
41.9
EUR
H61155.03
H61155.06
2-Brom-4-nitroimidazol ≥98%
2-Bromo-4-nitroimidazole
Formel:
C₃H₂BrN₃O₂ Molecular Weight: 191.97 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09025308 CAS-Nummer: 65902-59-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...