Methyl-5-fluor-2-nitrobenzoat ≥98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Fluor-2-nitrobenzoesäuremethylester

H61307.14 H61307.06
H61307.14EA 224 EUR
H61307.14 H61307.06
Methyl-5-fluor-2-nitrobenzoat ≥98%
Methyl-5-fluor-2-nitrobenzoat
Formel: C₈H₆FNO₄
Molecular Weight: 199,14 g/mol
Schmelzpunkt: 35…37 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03425619
CAS-Nummer: 393-85-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR