Cytochalasin D
Lieferant: Cayman Chemical
Synonyme:
7(S),18(R)-Dihydroxy-16(S),18-dimethyl-10-phenyl[11]cytochalasa-6(12),13(E),19(E)-trien-1,17-dion-21(R)-acetat
11330-1
11330-5
11330-10
CAYM11330-1EA
82
EUR
CAYM11330-1
CAYM11330-5
CAYM11330-10
Cytochalasin D
Cytochalasin D
Formel:
C₃₀H₃₇NO₆ Molecular Weight: 507.63 g/mol Schmelzpunkt: 259…260 °C |
MDL:
MFCD00077706 CAS-Nummer: 22144-77-0 EINECS: 244-804-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...