Colecalciferol (Vitamin D3)
Lieferant: Cayman Chemical
Synonyme: Calciol, (3β,5Z,7E)-9,10-Secocholesta- 5,7,10(19)-trien-3-ol, Vitamin D3, 3β,5-cis,7-trans-3-Hydroxy-9,10-secocholesta- 5,7,10(19)-trien, Colecalciferol, 3-[2-[7a-Methyl-1-(6-methylheptan-2-yl)- 2,3,3a,5,6,7-hexahydro-1H-inden-4-yliden] ethyliden]-4-methyliden-cyclohexan-1-ol
11792-1 11792-5 11792-10
CAYM11792-1EA 48 EUR
CAYM11792-1 CAYM11792-5 CAYM11792-10
Colecalciferol (Vitamin D3)
Colecalciferol (Vitamin D3)
Formel: C₂₇H₄₄O
Molecular Weight: 384.65 g/mol
MDL: MFCD00078131
CAS-Nummer: 67-97-0
EINECS: 200-673-2
UN: 2811
ADR: 6.1 II