N-Cyclohexylanilin
Lieferant: Thermo Scientific
SB01340DA
SB01340EA
SB01340.50
MAYBSB01340DAEA
0
EUR
MAYBSB01340DA
MAYBSB01340EA
MAYBSB01340.50
N-Cyclohexylanilin
N-Cyclohexylanilin
Formel:
C₁₂H₁₇N Molecular Weight: 175.27 g/mol Siedepunkt: 68 °C (0.001 mmHg) Schmelzpunkt: 16 Dichte: 1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014284 CAS-Nummer: 1821-36-9 EINECS: 217-344-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...