2-Brom-3-fluoranilin ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

H61687.03 H61687.06
H61687.03EA 106 EUR
H61687.03 H61687.06
2-Brom-3-fluoranilin ≥97%
2-Brom-3-fluoranilin

Purity: 97% CAS No.: 111721-75-6 Molecular Formula: C6H5BrFN Formula Weight: 190.01 Flash Point: 94°(201°F) Physical Form: Crystalline Density: 1.670 Refractive Index: 1.5830 Appearance: Off-white UN No.: 2811 MDL No.: MFCD07369915

Formel: C₆H₅BrFN
Molecular Weight: 190.01 g/mol
Dichte: 1.670
Flammpunkt: 94 °C (201 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD07369915
CAS-Nummer: 111721-75-6
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR