Serotonin Hydrochlorid

Lieferant: Cayman Chemical

Synonyme: 5-Hydroxytryptamine Hydrochloride

14332-50 14332-100
CAYM14332-50EA 50 EUR
CAYM14332-50 CAYM14332-100
Serotonin Hydrochlorid
Serotonin Hydrochlorid
Formel: C₁₀H₁₃ClN₂O
Molecular Weight: 212.68 g/mol
MDL: MFCD00012686
CAS-Nummer: 153-98-0
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR