2-(Methylamino)benzotrifluorid

Lieferant: Thermo Scientific

Synonyme: N-Methyl-2-(trifluoromethyl)aniline, 2-(Trifluoromethyl)-N-methylaniline

TL00239.1 TL00239.10 TL00239.50
MAYBTL00239.1EA 0 EUR
MAYBTL00239.1 MAYBTL00239.10 MAYBTL00239.50
2-(Methylamino)benzotrifluorid
2-(Methylamino)benzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175.15 g/mol
MDL: MFCD00052196
CAS-Nummer: 14925-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR