4-tert-Butylbenzonitril

Lieferant: Thermo Scientific

Synonyme: p-tert-Butylbenzonitril

TL00451.1 TL00451.10 TL00451.50
MAYBTL00451.1EA 0 EUR
MAYBTL00451.1 MAYBTL00451.10 MAYBTL00451.50
4-tert-Butylbenzonitril
4-tert-Butylbenzonitril
Formel: C₁₁H₁₃N
Molecular Weight: 159,23 g/mol
Siedepunkt: 67…69 °C (0,1 mmHg)
Dichte: 0,9492 g/cm³ (20 °C)
MDL: MFCD00075840
CAS-Nummer: 4210-32-6
EINECS: 224-137-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR