3-Fluorpicolinsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-fluoropyridine-2-carboxylic acid, 3-Fluoro-2-pyridinecarboxylic acid
H61458.03
H61458.06
H61458.03EA
77.2
EUR
H61458.03
H61458.06
3-Fluorpicolinsäure ≥98%
3-Fluorpicolinsäure
Formel:
C₆H₄FNO₂ Molecular Weight: 141,1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114108 CAS-Nummer: 152126-31-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...