(S)-(+)-2-Chlormandelsäure 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (S)-2-(2-Chlorophenyl)-2-hydroxyacetic acid

H61152.ME
H61152.MEEA 0 EUR
H61152.ME
(S)-(+)-2-Chlormandelsäure 95%
(S)-(+)-2-Chlormandelsäure
Formel: C₈H₇ClO₃
Molecular Weight: 186,59 g/mol
Schmelzpunkt: 118…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00798437
CAS-Nummer: 52950-19-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR