6,7-Dimethoxy-4-chinazolon
Lieferant: COMBI-BLOCKS
Synonyme:
6,7-Dimethoxy-4(3H)-quinazolinone, 6,7-Dimethoxy-1H-quinazolin-4-one
HC-6056-1G
HC-6056-25G
HC-6056-5G
COBBHC-6056-1GEA
25.5
EUR
COBBHC-6056-1G
COBBHC-6056-25G
COBBHC-6056-5G
6,7-Dimethoxy-4-chinazolon
6,7-Dimethoxy-4-chinazolon
Formel:
C₁₀H₁₀N₂O₃ Molecular Weight: 206,2 g/mol |
MDL:
MFCD01570147 CAS-Nummer: 13794-72-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...