Nitrofen

Lieferant: Thermo Scientific

Synonyme: 2,4-Dichloro-1-(4-nitrophenoxy)benzene

RDP00053DA RDP00053EA RDP00053.50
MAYBRDP00053DAEA 0 EUR
MAYBRDP00053DA MAYBRDP00053EA MAYBRDP00053.50
Nitrofen
Nitrofen
Formel: C₁₂H₇Cl₂NO₃
Molecular Weight: 284.1 g/mol
MDL: MFCD00128026
CAS-Nummer: 1836-75-5
EINECS: 217-406-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR