Streptozocin

Lieferant: Cayman Chemical
Gefahr

13104-50 13104-1
CAYM13104-50EA 32 EUR
CAYM13104-50 CAYM13104-1
Streptozocin
Streptozocin
Formel: C₈H₁₅N₃O₇
Lagertemperatur: Gefrierschrank
MDL: MFCD00006607
CAS-Nummer: 18883-66-4
EINECS: 242-646-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR