3'-Desoxyadenosin (Cordycepin)
Lieferant: Cayman Chemical
Synonyme: 3'-Deoxyadenosine, (2R,3R,5S)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3-ol, Cordycepin
14426-10 14426-25 14426-50 14426-100
CAYM14426-10EA 46 EUR
CAYM14426-10 CAYM14426-25 CAYM14426-50 CAYM14426-100
3'-Desoxyadenosin (Cordycepin)
3'-Desoxyadenosin (Cordycepin)
Formel: C₁₀H₁₃N₅O₃
Molecular Weight: 251.24 g/mol
MDL: MFCD00037998
CAS-Nummer: 73-03-0
EINECS: 200-791-4
UN: 2811
ADR: 6.1 III