5-Fluor-2'-deoxyuridin (Floxuridin)
Lieferant: Cayman Chemical
Synonyme: 5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine
14154-50 14154-100 14154-500
CAYM14154-50EA 68 EUR
CAYM14154-50 CAYM14154-100 CAYM14154-500
5-Fluor-2'-deoxyuridin (Floxuridin)
5-Fluor-2'-deoxyuridin (Floxuridin)
Formel: C₉H₁₁FN₂O₅
Molecular Weight: 246,19 g/mol
MDL: MFCD00006530
CAS-Nummer: 50-91-9
EINECS: 200-072-5
UN: 2811
ADR: 6.1,III