Ethyl-4-piperidincarboxylat
Lieferant: Thermo Scientific
Synonyme:
Ethyl piperidine-4-carboxylate, Ethyl isonipecotate, Ethyl 4-Piperidinecarboxylate
SB01810.1
SB01810.10
SB01810.50
MAYBSB01810.1EA
0
EUR
MAYBSB01810.1
MAYBSB01810.10
MAYBSB01810.50
Ethyl-4-piperidincarboxylat
Ethyl-4-piperidincarboxylat
Formel:
C₈H₁₅NO₂ Molecular Weight: 157,21 g/mol |
MDL:
MFCD00006003 CAS-Nummer: 1126-09-6 EINECS: 214-416-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...