2-Amino-4,5-dimethylthiazol Hydrochlorid 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4,5-Dimethylthiazol-2-amine hydrochloride

H59878.09 H59878.18
H59878.09EA 74.2 EUR
H59878.09 H59878.18
2-Amino-4,5-dimethylthiazol Hydrochlorid 98%
2-Amino-4,5-dimethylthiazol Hydrochlorid
Formel: C₅H₉ClN₂S
Molecular Weight: 164.66 g/mol
Schmelzpunkt: 273…274 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012711
CAS-Nummer: 71574-33-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR