2,6-Dichlornicotinonitril ≥97%

Lieferant: Thermo Scientific
Achtung

H63480.06EA 312 EUR
H63480.06 H63480.03
2,6-Dichlornicotinonitril ≥97%
2,6-Dichlornicotinonitril
Formel: C₆H₂Cl₂N₂
Molecular Weight: 173,00 g/mol
Schmelzpunkt: 121…125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03411341
CAS-Nummer: 40381-90-6
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR