α-Bromstyrol

Lieferant: COMBI-BLOCKS

Synonyme: alfa-Bromostyrene, α-Bromostyrene, alpha-Bromostyrene

OT-0670-5G OT-0670-1G
COBBOT-0670-5GEA 0 EUR
COBBOT-0670-5G COBBOT-0670-1G
α-Bromstyrol
α-Bromstyrol
Formel: C₈H₇Br
Molecular Weight: 183.0452 g/mol
MDL: MFCD00012229
CAS-Nummer: 98-81-7
EINECS: 202-702-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR