4-Methyl-3-nitrobenzotrifluorid
Lieferant: COMBI-BLOCKS
Synonyme:
1-Methyl-2-nitro-4-(trifluoromethyl)benzene
AN-1145-5G
AN-1145-25G
AN-1145-1G
COBBAN-1145-5GEA
88
EUR
COBBAN-1145-5G
COBBAN-1145-25G
COBBAN-1145-1G
4-Methyl-3-nitrobenzotrifluorid
4-Methyl-3-nitrobenzotrifluorid
Formel:
C₈H₆F₃NO₂ Molecular Weight: 205.1300 g/mol |
MDL:
MFCD00272402 CAS-Nummer: 65754-26-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...