Bis(α,α-dimethylbenzyl)peroxid (Dicumylperoxid) ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bis(1-methyl-1-phenylethyl)peroxide, Bis(alpha,alpha-dimethylbenzyl)peroxide, Dicumyl Peroxide
H60442.MC
H60442.36
H60442.22
H60442.MCEA
0
EUR
H60442.MC
H60442.36
H60442.22
Bis(α,α-dimethylbenzyl)peroxid (Dicumylperoxid) ≥98%
Bis(α,α-dimethylbenzyl)peroxid (Dicumylperoxid)
Formel:
C₁₈H₂₂O₂ Molecular Weight: 270.37 g/mol Schmelzpunkt: 39…41 °C Dichte: 1.56 Lagertemperatur: Kühlschrank |
MDL:
MFCD00036227 CAS-Nummer: 80-43-3 EINECS: 201-279-3 UN: 3110 ADR: 5.2 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...