trans-4-Aminocyclohexancarbonsäure Hydrochlorid

Lieferant: COMBI-BLOCKS

QA-5309-1G QA-5309-5G
COBBQA-5309-1GEA 0 EUR
COBBQA-5309-1G COBBQA-5309-5G
trans-4-Aminocyclohexancarbonsäure Hydrochlorid
trans-4-Aminocyclohexancarbonsäure Hydrochlorid
Formel: C₇H₁₄ClNO₂
MDL: MFCD03844587
CAS-Nummer: 27960-59-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR