Dropping bottles, APONORM®
Lieferant: MUELLER KREMPEL
14.015.10 14.015.12 14.015.13 14.015.14 14.015.11
MUEL14.015.10EA 15 EUR
MUEL14.015.10 MUEL14.015.12 MUEL14.015.13 MUEL14.015.14 MUEL14.015.11
Dropping bottles, APONORM®
Flaschen Tropfflaschen
Dropper bottles made of amber glass.
  • Good chemical and physical properties
  • For use in applications that do not involve high thermal stresses
  • Amber coloured to protect light sensitive substances
Thread according to DIN-18 and with dust protection plug, germ-free and hygienic shrink wrap packaging.