Safety storage depots for drums, SLH-NB
Lieferant: LACONT UMWELTTECHNIK
H61-1110-A H61-2110-A H61-1111-A H61-2111-A H61-1112-A
LACOH61-1110-AEA 0 EUR
LACOH61-1110-A LACOH61-2110-A LACOH61-1111-A LACOH61-2111-A LACOH61-1112-A
Safety storage depots for drums, SLH-NB
Möbel Lagerschränke
Safety storage depots are made of galvanised sheet steel with natural ventilation are approved for the passive storage of flammable liquids.
  • Reinforced roof and base profile frames
  • Circumferential splash protection made from sheet steel, except in door area
  • Built-in weather-protected ventilation slots in door and sidewalls
  • With ground connection for the avoidance of hazardous electrostatic charges
Base sump tray is made from 3 mm fire galvanised sheet steel, galvanised grid as support level.
The legally required 0,4-fold minimum air exchance rateMore Product Information