Combi top racks
Lieferant: KOEHLER TECHNISCHE PRODUKTEN
A-1182 A-1183
KOEHA-1182EA 34.3 EUR
KOEHA-1182 KOEHA-1183
Combi top racks
Gestelle Reagenzglasgestelle
Racks, made of polypropylene, neon coloured racks have four sides that can be used individually. with temperature resistant from −80 to 121 °C.
  • Temperature resistant from −80 to 121 °C.
  • Excellent chemical resistance
  • Sturdy