Alarm thermometer
Lieferant: TFA DOSTMANN
5020-0573 5020-0573Z
DOST5020-0573EA 70.4 EUR
DOST5020-0573 DOST5020-0573Z
Alarm thermometer
Thermometer Digitalthermometer
LT102 Alarm thermometer is designed for monitoring the temperature of liquid or semisolid material in laboratory refrigerators.
  • Water-tight
  • Big display
  • Celsius/Fahrenheit-switchable
The Hi-Lo-Alarm is the ideal function for detecting temperature problems during storage.
Lieferumfang: LT102 Alarm thermometer includes pocket clip, table rack, integrated sensor holder and magnet.
Bestellen
TECHNISCHE DATEN
Environmentally Preferable