Cutter for glass tubing
Lieferant: Hilgenberg
HILG4018303EA 134 EUR
HILG4018303 HILG4018304
Cutter for glass tubing
Schneidewerkzeuge
This cutter is suitable for cutting and scoring glass tubing.
  • Ceramic blades and aluminium holder
  • Suitable to cut off the bottom of Mark-tubes