L(+)-Asparagin, wasserfrei 98,0-102,0% für die biopharmazeutische Produktion

Lieferant: VWR Chemicals

Synonyme: (S)-(+)-Asparagin, L(+)-2-Amino-1,4-butandisäure-4-amid, (S)-(+)-2-Amino-1,4-butandisäure-4-amid

15005.297EA 1130 EUR
15005.297 15005.180 15005.366
L(+)-Asparagin, wasserfrei 98,0-102,0% für die biopharmazeutische Produktion
L(+)-Asparagin
Formel: H₂NCOCH₂CH(NH₂)CO₂H
Molecular Weight: 132,12 g/mol
Siedepunkt: 343 °C (1013 hPa)
Schmelzpunkt: 234…235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064401
CAS-Nummer: 70-47-3
EINECS: 200-735-9
Merck Index: 12,00872
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6f3bc8a15c3e973916b0a6ca62d12f6953e5a027&vid=3d09743d0834d64e4719e8295f634c39195c571f&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay 98.0 - 102.0 %
Appearance White crystalline powder
Cell culture toxicity Passes test
DNases Not detected
Identification by IR Conforms to structure
Proteases Not detected
RNases Not detected
Solubility (10 %; diluted HCl) Clear and complete
Spec. opt. rot. (C=10; HCl 6 N;on dried) 33.0 - 36.5 °
Aspartic acid Max. 2 %
Heavy metals (as Pb) Max. 15 ppm
Ignition residue Max. 0.1 %
Loss on drying (105°C; 3 h) Max. 0.5 %
Cl (Chloride) Max. 0.02 %
SO4 (Sulphate) Max. 0.03 %
Bioburden Max. 100 CFU/g
Endotoxin (2 g/l) Max. 0.04 EU/mg

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR