Crucibles with lids, alumina
Lieferant: Mettler - Toledo
00024123 51119960
METN00024123EA 824 EUR
METN00024123 METN51119960
Crucibles with lids, alumina
Tiegel
These reusable crucibles made from alumina (aluminum oxide) are used for TGA measurements. After mechanical cleaning, store them in water (or possibly even in 10% hydrochloric acid) because many salts are soluble in water. Oxides form chlorides in hydrochloric acid, which can be removed by rinsing. After drying, heat the crucibles to red heat to ensure that no weight loss occurs when they are used.
  • Withstand maximum temperature of 2000 °C
Bestellinformation: Special aluminum lids have to be ordered separately.