L(+)-Lysin Monohydrochlorid
Lieferant: G Bioscienes
Synonyme: L(+)-Lysin HydrochloridH-Lys-OH.HCl(S)-(+)-2,6-Diaminocapronsäure Hydrochlorid(S)-(+)-Lysin Monohydrochlorid(S)-(+)-Lysin Hydrochlorid(S)-(+)-2,6-Diaminohexansäure Hydrochlorid(S)-(+)-2,6-Diaminocapronsäure Monohydrochlorid(S)-(+)-2,6-Diaminohexansäure Monohydrochlorid
RC-066EA 77 EUR
RC-066 RC-067
L(+)-Lysin Monohydrochlorid
L(+)-Lysin Monohydrochlorid
Formel: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
Molecular Weight: 182,65 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00064564
CAS-Nummer: 657-27-2
EINECS: 213-091-9